Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
acrylic
feature
to hold 5 x single pipettes OR 2 x multipipettes
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio P4405
General description
Features and Benefits
- Universal – Fits all popular pipettes
- 5mm acrylic – 40% thicker
- Non-skid rubber feet
- Multi-channel “window” slots
- US Patent No 872,300
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description to hold, 5 x single pipettes OR 2 x multipipettes | description to hold, 6 x single pipettes OR 4 x multipipettes | description to hold, 8 x single pipettes OR 6 x multipipettes | description to hold, 6 x Halo™ single pipettes OR 6 x ProPette™ LE single pipettes OR 4 x ProPette™multipipettes |
| feature to hold 5 x single pipettes OR 2 x multipipettes | feature to hold 6 x single pipettes OR 4 x multipipettes | feature to hold 8 x single pipettes OR 6 x multipipettes | feature to hold 6 x Halo™ single pipettes OR 6 x ProPette™ LE single pipettes OR 4 x ProPette™multipipettes |
| manufacturer/tradename MTC™ Bio P4405 | manufacturer/tradename MTC™ Bio P4406 | manufacturer/tradename MTC™ Bio P4408 | manufacturer/tradename MTC™ Bio P4406-P |
| material acrylic | material acrylic | material acrylic | material acrylic |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under MTC™ Bio SureStand™ Multiple Pipette Stand
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?