Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2470,00 zł 2099,50 zł |
Informacje o tej pozycji
2099,50 zł
Cena katalogowa2470,00 złZaoszczędź 15%material
plastic , silicone
feature
CE compliant
manufacturer/tradename
Drummond Scientific 4-000-201
volume
1-100 mL
General description
- Pipet-Aid® automatic pipette is of light-weight and compact design.
- Three-speed range options (F) Fast, (M) Medium, (S) Slow provide more precise control for aspirating and dispensing samples.
- EZ Grip nosepiece with double membrane filter.
- Includes a removable stand.
- Easy access battery replacement.
- Holster-type wall bracket included.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description model XP, cordless, rechargeable, AC/DC input 220 V AC, European 2-pin plug | description cordless, rechargeable, AC/DC input 220 V AC, European 2-pin plug | description model XP, cordless, rechargeable, AC/DC input 110 V AC, US 2-pin plug | description cordless, rechargeable, AC/DC input 110 V AC |
| volume 1-100 mL | volume 1-100 mL | volume 1-100 mL | volume 1-100 mL |
| feature CE compliant | feature CE compliant | feature - | feature - |
| manufacturer/tradename Drummond Scientific 4-000-201 | manufacturer/tradename Drummond Scientific 4-000-200 | manufacturer/tradename Drummond Scientific 4-000-101 | manufacturer/tradename Drummond Scientific 4-000-100 |
| material plastic , silicone | material silicone , plastic | material plastic , silicone | material plastic , silicone |
Still not finding the right product?
Explore all of our products under Pipet-Aid® automatic pipette pumps
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?