Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
plastic , silicone
feature
CE compliant
manufacturer/tradename
Drummond Scientific 4-000-200
volume
1-100 mL
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description cordless, rechargeable, AC/DC input 220 V AC, European 2-pin plug | description model XP, cordless, rechargeable, AC/DC input 110 V AC, US 2-pin plug | description model XP, cordless, rechargeable, AC/DC input 220 V AC, European 2-pin plug | description cordless, rechargeable, AC/DC input 110 V AC |
| volume 1-100 mL | volume 1-100 mL | volume 1-100 mL | volume 1-100 mL |
| feature CE compliant | feature - | feature CE compliant | feature - |
| manufacturer/tradename Drummond Scientific 4-000-200 | manufacturer/tradename Drummond Scientific 4-000-101 | manufacturer/tradename Drummond Scientific 4-000-201 | manufacturer/tradename Drummond Scientific 4-000-100 |
| material silicone , plastic | material plastic , silicone | material plastic , silicone | material plastic , silicone |
Still not finding the right product?
Explore all of our products under Pipet-Aid® automatic pipette pumps
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?