Przejdź do
S6690
Nunc® Lab-Tek® II - CC2™ Chamber Slide™ system
4 wells, glass slide, 1.7 cm2/well, sterile
Synonim(y):
cell culture chamber, chamber slides, microscope slides, microscopy slides, Nunc 154917
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122600
Material:
colorless , polystyrene chamber
Sterility:
sterile
Packaging:
pkg of carton of 16
Wells:
4
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
colorless , polystyrene chamber
sterility
sterile
feature
CE marked
manufacturer/tradename
Nunc 154917
packaging
pkg of carton of 16
technique(s)
cell culture: suitable
well area
1.7 cm2
wells
4
working volume
0.5-1.0 mL
suitability
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
General description
Nunc® Lab-Tek® II - CC2 Chamber Slide System
  • Chemically coated growth surface on glass slide that mimics polylysine
  • Provides binding sites optimal for fastidious cells (e.g. neurons)
  • Growth surface remains stable without refrigeration
  • Light blue frosted writing area
  • Slide seperator included in each pack
  • CE marked
Legal Information
CC2 is a trademark of Thermo Fisher Scientific or its subsidiaries
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
S6815S6565C7057
description
4 wells, glass slide, 1.7 cm2/well, sterile
description
8 wells, glass slide, 0.7 cm2/well, sterile
description
2 wells, glass slide, 4.0 cm2/well, sterile
description
wells 8, well area 0.7 cm2, sterile
manufacturer/tradename
Nunc 154917
manufacturer/tradename
Nunc 154941
manufacturer/tradename
Nunc 154852
manufacturer/tradename
Nunc 154534
material
polystyrene chamber, colorless
material
polystyrene chamber, colorless
material
polystyrene chamber
material
polystyrene chamber
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
wells
4
wells
8
wells
-
wells
8
packaging
pkg of carton of 16
packaging
pkg of carton of 16
packaging
pkg of carton of 16
packaging
pack of 16 ×
working volume
0.5-1.0 mL
working volume
0.2-0.5 mL
working volume
1.0-2.0 mL
working volume
0.2-0.5 mL
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)