Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
colorless , polystyrene chamber
sterility
sterile
feature
CE marked
manufacturer/tradename
Nunc 154941
packaging
pkg of carton of 16
technique(s)
cell culture: suitable
well area
0.7 cm2
wells
8
working volume
0.2-0.5 mL
suitability
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
General description
- Chemically coated growth surface on glass slide that mimics polylysine
- Provides binding sites optimal for fastidious cells (e.g. neurons)
- Growth surface remains stable without refrigeration
- Light blue frosted writing area
- Slide seperator included in each pack
- CE marked
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 8 wells, glass slide, 0.7 cm2/well, sterile | description 4 wells, glass slide, 1.7 cm2/well, sterile | description 2 wells, glass slide, 4.0 cm2/well, sterile | description wells 8, well area 0.7 cm2, sterile |
| manufacturer/tradename Nunc 154941 | manufacturer/tradename Nunc 154917 | manufacturer/tradename Nunc 154852 | manufacturer/tradename Nunc 154534 |
| material polystyrene chamber, colorless | material polystyrene chamber, colorless | material polystyrene chamber | material polystyrene chamber |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| wells 8 | wells 4 | wells - | wells 8 |
| packaging pkg of carton of 16 | packaging pkg of carton of 16 | packaging pkg of carton of 16 | packaging pack of 16 × |
| well area 0.7 cm2 | well area 1.7 cm2 | well area - | well area 0.7 cm2 |
Still not finding the right product?
Explore all of our products under Nunc® Lab-Tek® II - CC2™ Chamber Slide™ system
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?