Przejdź do zawartości
Merck

Przejdź do

S9655

Active Recombinant Human Superoxide Dismutase

Lyophilized powder, >20,000U/mg protein

Synonim(y):

SOD1, superoxide dismutase 1, Cu/Zn superoxide dismutase, 5-LO, Cu/Zn SOD, Human, Recombinant, E. coli

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o tej pozycji

UNSPSC Code:
12352204
Form:
lyophilized powder
Biological source:
human
Mol wt:
16 kDa
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


biological source

human

Quality Segment

form

lyophilized powder

specific activity

≥20000 U/mg

mol wt

16 kDa

purified by

chromatography

suitability

suitable for molecular biology

UniProt accession no.

shipped in

wet ice

storage temp.

-10 to -25°C

Gene Information

human ... SOD1(6647)

General description

Superoxide Dismutase is a metalloenzyme that catalyzes the dismutation of superoxide radicals into molecular oxygen and hydrogen peroxide. Multiple isoforms exist in mammalian systems, including cytosolic Cu/Zn-SOD (SOD1), mitochondrial Mn-SOD (SOD2), and extracellular SOD (SOD3), each differing in metal cofactor and subcellular localization. SOD plays a fundamental role in cellular defense against oxidative stress and is essential for maintaining redox balance.

Application

Superoxide dismutase has been used in:
  • Antioxidant activity assays
  • Oxidative stress and ROS studies
  • Cell protection and apoptosis studies
  • Drug screening and toxicity studies

Biochem/physiol Actions

SOD catalyzes the conversion of superoxide anions (O₂⁻) into hydrogen peroxide (H₂O₂) and molecular oxygen, thereby protecting cells from oxidative damage. It plays an essential role in maintaining redox homeostasis and preventing cellular injury.

Physical form

20mM PB (pH 7.0), 0.1M NaCl
Centrifuge the vial at 4ºC before opening to recover the entire contents. It is recommended that sterile water be added to the vial to prepare a stock solution.

Preparation Note

Centrifuge the vial at 4ºC before opening to recover the entire contents. It is recommended that sterile water be added to the vial to prepare a stock solution.
Unit definition: One unit of SOD is defined as the amount of enzyme causing half the maximum inhibition of the oxidation of 7.5 mM NADH in the presence of EDTA, manganese ions, and mercaptoethanol at 23°C and pH 7.4 in lnvitrogen Superoxide Dismutase (SOD) Colorimetric Activity Kit (Cat# EIASODC)
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
SAB4200807ZRB1508574591
description

Lyophilized powder, >20,000U/mg protein

description

clone SD-G6, purified from hybridoma cell culture

description

recombinant, expressed in HEK 293 cells

description

-

biological source

human

biological source

mouse

biological source

rabbit

biological source

-

suitability

suitable for molecular biology

suitability

-

suitability

-

suitability

-

form

lyophilized powder

form

buffered aqueous solution

form

lyophilized

form

solid

mol wt

16 kDa

mol wt

-

mol wt

observed mol wt ~16 kDa, calculated mol wt 15.94 kDa

mol wt

-

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

specific activity

≥20000 U/mg

specific activity

-

specific activity

-

specific activity

≥3000 U/mg


Piktogramy

Health hazard

Słowo ostrzegawcze

Danger

Kody zagrożeń

Ostrzeżenia

Hazard Classifications

Resp. Sens. 1

Klasa składowania

11 - Combustible Solids



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów