Przejdź do zawartości
Merck

Przejdź do

SML4305

Ceftiofur sodium salt

≥98% (HPLC), powder, antibiotic

Synonim(y):

(6R,7R)-7-[[(2Z)-(2-Amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[(2-furanylcarbonyl)thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monosodium salt, Sodium (6R,7R)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-(((furan-2-carbonyl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Ceftiofur sodium, U 64279E

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
537,00 zł

Informacje o tej pozycji

Wzór empiryczny (zapis Hilla):
C19H16N5NaO7S3
Numer CAS:
Masa cząsteczkowa:
545.54
MDL number:
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

537,00 zł


Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności

​
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

General description

Ceftiofur sodium salt, a third-generation β-lactam antibiotic, exhibits broad-spectrum bactericidal activity against many veterinary pathogens.
potent, broad-spectrum β-lactam bactericidally inhibits bacterial cell wall synthesis via PBP binding, its active metabolite boosting veterinary efficacy and ensuring safety.

Ceftiofur sodium salt a third-generation β-lactam antibiotic, exhibits broad-spectrum bactericidal activity against many veterinary pathogens. Its core mechanism disrupts bacterial cell wall synthesis by irreversibly inactivating penicillin-binding proteins (PBPs) on the inner bacterial membrane, causing cell lysis. It shows potent activity with low MIC values against key pathogens like Pasteurella multocida (MIC90 ≤ 0.03 µg/ml) and Actinobacillus pleuropneumoniae (MIC90 ≤ 0.125 µg/ml). Parenterally administered due to poor oral absorption, ceftiofur rapidly metabolizes in vivo to desfuroylceftiofur (DFC), an equally active metabolite enhancing its efficacy. With a good therapeutic safety profile, ceftiofur is a significant veterinary antimicrobial for susceptible infections.

Application

Ceftiofur sodium salt may be used in the assessment of biodegradation mechanisms of ceftiofur sodium by Escherichia sp. CS-1.

Biochem/physiol Actions

Ceftiofur sodium salt disrupts bacterial cell wall synthesis by irreversibly inactivating penicillin-binding proteins (PBPs) on the inner bacterial membrane, causing cell lysis. It shows potent activity with low MIC values against key pathogens like Pasteurella multocida (MIC90 ≤ 0.03 µg/ml) and Actinobacillus pleuropneumoniae (MIC90 ≤ 0.125 µg/ml). Parenterally administered due to poor oral absorption, ceftiofur rapidly metabolizes in vivo to desfuroylceftiofur (DFC), an equally active metabolite enhancing its efficacy. With a good therapeutic safety profile, ceftiofur is a significant veterinary antimicrobial for susceptible infections.

Disclaimer

Hygroscopic Store with desiccant
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
PHR1521109807132422
description

≥98% (HPLC), powder, antibiotic

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

United States Pharmacopeia (USP) Reference Standard

description

VETRANAL®, analytical standard

form

powder

form

-

form

-

form

-

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

100

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

-

solubility

H2O: 2 mg/mL, clear (warmed)

solubility

-

solubility

-

solubility

-

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-


Piktogramy

Health hazard

Słowo ostrzegawcze

Danger

Kody zagrożeń

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Klasa składowania

11 - Combustible Solids

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów