Przejdź do
T2543
Nalgene® centrifuge tubes, Oak Ridge Style 3139
nominal capacity 10 mL
Synonim(y):
High-Speed ​​Centrifuge Tubes, Nalgene tubes, Nalgene 3139-0010, sample collection tube, sample receptacle, sample tube
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.11
UNSPSC Code:
41121703
eCl@ss:
32040302
Material:
polypropylene copolymer
Packaging:
pack of 10 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene copolymer
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3139-0010
nominal capacity
10 mL
General description
Spin with confidence with when you use these Nalgene® Oak Ridge high-speed PPCO centrifuge tubes complete with sealing caps. These tubes are chemically compatible with a wider variety of laboratory reagents than polycarbonate centrifuge tubes and are repeately autoclavable for extended performance.
Features and Benefits
  • Greater chemical resistance than polycarbonate
  • Autoclavable
  • Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
  • 3139-0050 (T2918) can be used up to 100,605 x g in the T29-8x50 rotor
  • Comes with polypropylene sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
  • Must be filled to 80% of total capacity when centrifuging
  • Autoclavable/Leakproof† when used with supplied Nalgene closure; Repeated autoclaving will reduce the life of PC centrifuge tubes.
Packaging
With sealing cap
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
T2793T2668T2918
description
nominal capacity 10 mL
description
nominal capacity 30 mL
description
nominal capacity 16 mL
description
nominal capacity 50 mL
manufacturer/tradename
Nalgene® 3139-0010
manufacturer/tradename
Nalgene® 3139-0030
manufacturer/tradename
Nalgene® 3139-0016
manufacturer/tradename
Nalgene® 3139-0050
material
polypropylene copolymer
material
cap (Sealing cap), polypropylene , round bottom tube, polypropylene copolymer
material
polypropylene , round bottom tube, cap (Sealing cap), polypropylene copolymer
material
cap (Sealing cap), polypropylene copolymer
packaging
pack of 10 ea
packaging
pack of 10 ea
packaging
pack of 10 ea
packaging
pack of 10 ea
nominal capacity
10 mL
nominal capacity
30 mL
nominal capacity
16 mL
nominal capacity
50 mL
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)