Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 877,00 zł |
Informacje o tej pozycji
877,00 zł
material
cap (Sealing cap), polypropylene , polypropylene copolymer , round bottom tube
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3139-0016
nominal capacity
16 mL
General description
Features and Benefits
- Greater chemical resistance than polycarbonate
- Autoclavable
- Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges
- 3139-0050 (T2918) can be used up to 100,605 x g in the T29-8x50 rotor
- Comes with polypropylene sealing caps with silicone gaskets for extra leak protection when spinning >10,000 x g or spinning hazardous materials
- Must be filled to 80% of total capacity when centrifuging
- Autoclavable/Leakproof† when used with supplied Nalgene closure; Repeated autoclaving will reduce the life of PC centrifuge tubes.
Packaging
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description nominal capacity 16 mL | description nominal capacity 30 mL | description nominal capacity 10 mL | description nominal capacity 50 mL |
| manufacturer/tradename Nalgene® 3139-0016 | manufacturer/tradename Nalgene® 3139-0030 | manufacturer/tradename Nalgene® 3139-0010 | manufacturer/tradename Nalgene® 3139-0050 |
| sterility non-sterile | sterility - | sterility - | sterility non-sterile |
| material polypropylene copolymer , cap (Sealing cap), round bottom tube | material polypropylene , cap (Sealing cap), round bottom tube, polypropylene copolymer | material polypropylene copolymer | material cap (Sealing cap), polypropylene copolymer |
| packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea | packaging pack of 10 ea |
| feature RCF 50,000 × g | feature RCF 50,000 × g | feature - | feature RCF 100,605 × g |
| nominal capacity 16 mL | nominal capacity 30 mL | nominal capacity 10 mL | nominal capacity 50 mL |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge tubes, Oak Ridge Style 3139
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?