Przejdź do
TMO150200
Nunc® Non-Treated Multidish
wells 12, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea
Synonim(y):
multiwell plate, non treated multiwell plate, non treated plate, Nunc 150200, cultivation plate
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.13
UNSPSC Code:
41161502
eCl@ss:
32049090
Material:
polystyrene
Manufacturer/tradename:
Nunc 150200
Packaging:
case of 75 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polystyrene
packaging
case of 75 ea
manufacturer/tradename
Nunc 150200
technique(s)
cell culture: suitable
L × W
128 mm × 86 mm
wells
12
General description
Perform suspension cell cultures with Thermo Scientific Nunc Non-Treated Multidishes.
Application
  • Suspension cell cultures.
Features and Benefits
  • Flat-bottom round wells.
  • Excellent optical properties.
  • Raised well rims to lower the risk of cross-contamination.
  • Lid with notched corners for good fit and easy orientation.
  • Sterile and non-pyrogenic.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
TMO150787TMO150239TMO144530
description
wells 12, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea
description
wells 48, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea
description
wells 6, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea
description
wells 24, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea
manufacturer/tradename
Nunc 150200
manufacturer/tradename
Nunc 150787
manufacturer/tradename
Nunc 150239
manufacturer/tradename
Nunc 144530
material
polystyrene
material
polystyrene
material
polystyrene
material
polystyrene
packaging
case of 75 ea
packaging
case of 75 ea
packaging
case of 75 ea
packaging
case of 75 ea
L × W
128 mm × 86 mm
L × W
128 mm × 86 mm
L × W
128 mm × 86 mm
L × W
128 mm × 86 mm
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
technique(s)
cell culture: suitable
wells
12
wells
48
wells
6
wells
24
Still not finding the right product?
Explore all of our products under Nunc® Non-Treated Multidish
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)