Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
polystyrene
packaging
case of 75 ea
manufacturer/tradename
Nunc 150239
technique(s)
cell culture: suitable
L × W
128 mm × 86 mm
wells
6
General description
Application
- Suspension cell cultures.
Features and Benefits
- Flat-bottom round wells.
- Excellent optical properties.
- Raised well rims to lower the risk of cross-contamination.
- Lid with notched corners for good fit and easy orientation.
- Sterile and non-pyrogenic.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description wells 6, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea | description wells 24, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea | description wells 48, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea | description wells 12, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea |
| manufacturer/tradename Nunc 150239 | manufacturer/tradename Nunc 144530 | manufacturer/tradename Nunc 150787 | manufacturer/tradename Nunc 150200 |
| material polystyrene | material polystyrene | material polystyrene | material polystyrene |
| L × W 128 mm × 86 mm | L × W 128 mm × 86 mm | L × W 128 mm × 86 mm | L × W 128 mm × 86 mm |
| packaging case of 75 ea | packaging case of 75 ea | packaging case of 75 ea | packaging case of 75 ea |
| technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable |
| wells 6 | wells 24 | wells 48 | wells 12 |
Still not finding the right product?
Explore all of our products under Nunc® Non-Treated Multidish
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?