Przejdź do zawartości
Merck

Przejdź do

Z-2103

ZytoLight® SPEC CDK4/CEN 12 Dual Color Probe

green to orange, pkg of 0.05 mL, pkg of 20 tests

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


product line

ZytoVision

packaging

pkg of 0.05 mL, pkg of 0.2 mL, pkg of 20 tests, pkg of 5 tests

manufacturer/tradename

ZytoVision

storage condition

protect from light

color

green to orange

application(s)

research use

storage temp.

2-8°C

General description

The ZytoLight SPEC CDK4/CEN 12 Dual Color Probe (PL61) is a direct-labeled FISH probe designed for the qualitative detection of amplifications involving the human CDK4 gene and for detecting specific sequences on chromosome 12 in formalin-fixed, paraffin-embedded (FFPE) specimens. It is intended to be used in combination with the ZytoLight FISH-Tissue Implementation Kit.

Principle:

The fluorescence in situ hybridization (FISH) technique detects and visualizes specific nucleic acid sequences using fluorescently-labeled DNA fragments (FISH probes) that co-denature and hybridize with complementary target DNA strands. After stringent washing to remove unbound fragments, the hybridized probes are visualized with a fluorescence microscope utilizing specific excitation and emission filters.In this method, fluorescently labeled locus-specific (CDK4) and centromeric (CEN 12) DNA probes co-denature with target DNA in pretreated formalin-fixed, paraffin-embedded (FFPE) sections (10 µl of probe per slide, sealed with a coverslip and rubber cement). Following denaturation at 75°C for 10 minutes and overnight hybridization at 37°C, stringent washing and DAPI counterstaining enable visualization with color-specific filters according to the ZytoLight FISH-Tissue Implementation Kit protocol.

Components:

ZytoLight® SPEC CDK4/CEN 12 Dual Color Probe comprises:

  • ZyGreen labeled polynucleotides (excitation 503 nm/emission 528 nm) targeting the CDK4 gene region on chromosome 12 (12q13.3).
  • ZyOrange labeled polynucleotides (excitation 547 nm/emission 572 nm) targeting the alpha satellite centromeric region D12Z3 of chromosome 12.
  • Formamide-based hybridization buffer.

Application

The probe set enables qualitative detection of the CDK4 gene region and chromosome 12 centromere in formalin-fixed, paraffin-embedded (FFPE) tissue specimens by FISH. Used with ZytoLight® FISH-Tissue Implementation Kit for specimen pretreatment (dewaxing, proteolysis), denaturation/hybridization, and post-hybridization processing (washing, counterstaining).

Analysis Note

In a normal interphase nucleus, two green signals (for CDK4) and two orange signals (for CEN 12) are expected. Amplifications of the CDK4 gene region will result in an increased number of green signals or clusters. Any unexpected signal patterns should be investigated for potential genetic rearrangements.

Legal Information

ZytoLight is a registered trademark of ZytoVision GmbH in Germany
Ta strona może zawierać tekst przetłumaczony maszynowo.


Piktogramy

Health hazard

Słowo ostrzegawcze

Danger

Kody zagrożeń

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Narządy docelowe

Blood

Klasa składowania

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Wykazy regulacyjne

Wykazy regulacyjne dotyczą głównie produktów chemicznych. Można w nich podawać ograniczoną liczbę informacji na temat produktów niechemicznych. Brak wpisu oznacza, że żaden ze składników nie znajduje się w wykazie. Użytkownik odpowiada za zagwarantowanie bezpiecznego i zgodnego z prawem stosowania produktu.

75-12-7

CAS No.

75-12-7

CAS No.


Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów