Przejdź do
1A13070
USP
2,5-Thiazolylmethyldicarbamate
Pharmaceutical Analytical Impurity (PAI)
Synonim(y):
2,5-Thiazolylmethyldicarbamate
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
grade
pharmaceutical analytical impurity (PAI)
agency
USP
manufacturer/tradename
USP
application(s)
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the issuing Pharmacopoeia.">website
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
USP issued SDS can be found here">here
Piktogramy
Exclamation mark
Słowo ostrzegawcze
Warning
Kody zagrożeń
Hazard Classifications
Eye Irrit. 2
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)