Merck
Vasopressin-secreting neurons derived from human embryonic stem cells through specific induction of dorsal hypothalamic progenitors.
Scientific reports (2018-02-28)
Koichiro Ogawa, Hidetaka Suga, Chikafumi Ozone, Mayu Sakakibara, Tomiko Yamada, Mayuko Kano, Kazuki Mitsumoto, Takatoshi Kasai, Yu Kodani, Hiroshi Nagasaki, Naoki Yamamoto, Daisuke Hagiwara, Motomitsu Goto, Ryoichi Banno, Yoshihisa Sugimura, Hiroshi Arima
ABSTRAKT
Arginine-vasopressin (AVP) neurons exist in the hypothalamus, a major region of the diencephalon, and play an essential role in water balance. Here, we established the differentiation method for AVP-secreting neurons from human embryonic stem cells (hESCs) by recapitulating in vitro the in vivo embryonic developmental processes of AVP neurons. At first, the differentiation efficiency was improved. That was achieved through the optimization of the culture condition for obtaining dorsal hypothalamic progenitors. Secondly, the induced AVP neurons were identified by immunohistochemistry and these neurons secreted AVP after potassium chloride stimulation. Additionally, other hypothalamic neuropeptides were also detected, such as oxytocin, corticotropin-releasing hormone, thyrotropin-releasing hormone, pro-opiomelanocortin, agouti-related peptide, orexin, and melanin-concentrating hormone. This is the first report describing the generation of secretory AVP neurons derived from hESCs. This method will be applicable to research using disease models and, potentially, for regenerative medicine of the hypothalamus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-MCH antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Inhibitor Akt VIII, izoenzymoselektywny, Akti-1/2, Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2, CAS 612847-09-3, is a cell-permeable, reversible inhibitor of Akt1/Akt2 (IC50 = 58 nM, 210 nM, & 2.12 µM for Akt1, Akt2, and Akt3, respectively).
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Oxytocin Antibody, clone 4G11, ascites fluid, clone 4G11, Chemicon®
Informacje o cenach i dostępności nie są obecnie dostępne.