Merck
Macropinocytosis of Nab-paclitaxel Drives Macrophage Activation in Pancreatic Cancer.
Cancer immunology research (2017-01-22)
Jane Cullis, Despina Siolas, Antonina Avanzi, Sugata Barui, Anirban Maitra, Dafna Bar-Sagi
ABSTRAKT
Pancreatic cancer is a devastating disease that is largely refractory to currently available treatment strategies. Therapeutic resistance is partially attributed to the dense stromal reaction of pancreatic ductal adenocarcinoma tumors that includes a pervasive infiltration of immunosuppressive (M2) macrophages. Nab-paclitaxel (trade name Abraxane) is a nanoparticle albumin-bound formulation of paclitaxel that, in combination with gemcitabine, is currently the first-line treatment for pancreatic cancer. Here, we show that macrophages internalized nab-paclitaxel via macropinocytosis. The macropinocytic uptake of nab-paclitaxel induced macrophage immunostimulatory (M1) cytokine expression and synergized with IFNγ to promote inducible nitric oxide synthase expression in a TLR4-dependent manner. Nab-paclitaxel was internalized by tumor-associated macrophages in vivo, and therapeutic doses of nab-paclitaxel alone, and in combination with gemcitabine, increased the MHCII+CD80+CD86+ M1 macrophage population. These data revealed an unanticipated role for nab-paclitaxel in macrophage activation and rationalized its potential use to target immune evasion in pancreatic cancer. Cancer Immunol Res; 5(3); 182-90. ©2017 AACR.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Informacje o cenach i dostępności nie są obecnie dostępne.