- FKBP25, a novel regulator of the p53 pathway, induces the degradation of MDM2 and activation of p53.
FKBP25, a novel regulator of the p53 pathway, induces the degradation of MDM2 and activation of p53.
FEBS letters (2009-01-27)
Anna Maria Ochocka, Petros Kampanis, Samantha Nicol, Nerea Allende-Vega, Miranda Cox, Lynnette Marcar, Diane Milne, Frances Fuller-Pace, David Meek
PMID19166840
ABSTRAKT
The p53 tumour suppressor protein is tightly controlled by the E3 ubiquitin ligase, mouse double minute 2 (MDM2), but maintains MDM2 expression as part of a negative feedback loop. We have identified the immunophilin, 25kDa FK506-binding protein (FKBP25), previously shown to be regulated by p53-mediated repression, as an MDM2-interacting partner. We show that FKBP25 stimulates auto-ubiquitylation and proteasomal degradation of MDM2, leading to the induction of p53. Depletion of FKBP25 by siRNA leads to increased levels of MDM2 and a corresponding reduction in p53 and p21 levels. These data are consistent with the idea that FKBP25 contributes to regulation of the p53-MDM2 negative feedback loop.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciwko acetylo-lizynie, klon 4G12, clone 4G12, Upstate®, from mouse
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution