Merck
IL-33 promotes gastrointestinal allergy in a TSLP-independent manner.
Mucosal immunology (2017-06-29)
H Han, F Roan, L K Johnston, D E Smith, P J Bryce, S F Ziegler
ABSTRAKT
Atopic dermatitis (AD) often precedes asthma and food allergy, indicating that epicutaneous sensitization to allergens may be important in the induction of allergic responses at other barrier surfaces. Thymic stromal lymphopoietin (TSLP) and interleukin (IL)-33 are two cytokines that may drive type 2 responses in the skin; both are potential targets in the treatment of allergic diseases. We tested the functional role of IL-33 and the interplay between IL-33 and TSLP in mouse models of atopic march and gastrointestinal (GI) allergy. IL-33-driven allergic disease occurred in a TSLP-independent manner. In contrast, mice lacking IL-33 signaling were protected from onset of allergic diarrhea in TSLP-driven disease. Epithelial-derived IL-33 was important in this model, as specific loss of IL-33 expression in the epithelium attenuated cutaneous inflammation. Notably, the development of diarrhea following sensitization with TLSP plus antigen was ameliorated even when IL-33 was blocked after sensitization. Thus, IL-33 has an important role during early cutaneous inflammation and during challenge. These data reveal critical roles for IL-33 in the "atopic march" that leads from AD to GI allergy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Supelco
Albumin from chicken egg white, For use as a marker in SDS-PAGE