Merck
Casein kinase 2 phosphorylates and stabilizes C/EBPβ in pancreatic β cells.
Biochemical and biophysical research communications (2018-02-16)
Tomoko Takai, Tomokazu Matsuda, Yuki Matsuura, Kaho Inoue, Emi Suzuki, Ayumi Kanno, Maki Kimura-Koyanagi, Shun-Ichiro Asahara, Naoya Hatano, Wataru Ogawa, Yoshiaki Kido
ABSTRAKT
During the development of type 2 diabetes, endoplasmic reticulum (ER) stress leads to pancreatic β cell failure. CCAAT/enhancer-binding protein (C/EBP) β is highly induced by ER stress and AMP-activated protein kinase (AMPK) suppression in pancreatic β cells, and its accumulation reduces pancreatic β cell mass. We investigated the phosphorylation state of C/EBPβ under these conditions. Casein kinase 2 (CK2) was found to co-localize with C/EBPβ in MIN6 cells. It phosphorylated S222 of C/EBPβ, a previously unidentified phosphorylation site. We found that C/EBPβ is phosphorylated by CK2 under AMPK suppression and ER stress, which are important from the viewpoint of the worsening pathological condition of type 2 diabetes, such as decreased insulin secretion and apoptosis of pancreatic β cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.