Merck
Novel secondary mutations C56S and G149A confer resistance to HIV-1 integrase strand transfer inhibitors.
Antiviral research (2018-02-08)
Tomokazu Yoshinaga, Takahiro Seki, Shigeru Miki, Tadashi Miyamoto, Akemi Suyama-Kagitani, Shinobu Kawauchi-Miki, Masanori Kobayashi, Akihiko Sato, Eugene Stewart, Mark Underwood, Tamio Fujiwara
ABSTRAKT
Cabotegravir (CAB, S/GSK1265744) is an investigational second-generation integrase strand transfer inhibitor (INSTI) with a chemical structure similar to dolutegravir. CAB is under development as a long-acting injectable formulation for treatment of HIV-1 infection and for pre-exposure prophylaxis. We conducted an in vitro passage study of raltegravir- or elvitegravir-resistant signature mutants in the presence of CAB to characterize the resistance profile of this drug. During passage with Q148H virus, G140S arose by day 14, followed by G149A and C56S. Using site-directed mutagenesis, we obtained HIV molecular clones containing mutations encoding C56S and G149A in the integrase-coding region. Those substitutions were characterized in vitro as INSTI-resistance-associated secondary resistance mutations. Signature mutant viruses G140S/Q148H in which C56S and G149A were added acquired further INSTI resistance in conjunction with diminished integration activity, which yielded slower growth under drug-free conditions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Di-o-tolylguanidine, 99%
Informacje o cenach i dostępności nie są obecnie dostępne.