Merck
  • MicroRNA-421 suppresses the apoptosis and autophagy of hippocampal neurons in epilepsy mice model by inhibition of the TLR/MYD88 pathway.
MicroRNA-421 suppresses the apoptosis and autophagy of hippocampal neurons in epilepsy mice model by inhibition of the TLR/MYD88 pathway.
Journal of cellular physiology (2018-01-31)
Xin Wen, Xin-Rui Han, Yong-Jian Wang, Shan Wang, Min Shen, Zi-Feng Zhang, Shao-Hua Fan, Qun Shan, Liang Wang, Meng-Qiu Li, Bin Hu, Chun-Hui Sun, Dong-Mei Wu, Jun Lu, Yuan-Lin Zheng
ABSTRAKT
Epilepsy is a group of neurological disorders characterized by epileptic seizures. In this study, we aim to explore the role of microRNA-421 (miR-421) in hippocampal neurons of epilepsy mice via the TLR/MYD88 pathway. Forty mice were randomly served as the normal and model (established as epilepsy model) groups. Hippocampal neurons were assigned into seven groups with different transfections. The RT-qPCR and western blotting were conducted to examine the expression of miR-421 TLR2, TLR4, MYD88, Bax, Bcl-2, p53, Beclin-1, and LC3II/LC3I. Cell proliferation and apoptosis were detected by MTT and flow cytometry.MYD88 is a target gene of miR-421. Model mice showed elevated expression of TLR2, TLR4, MYD88, Bax, p53, Beclin-1, and LC3II/LC3I but reduced expression of miR-421 and Bcl-2. In vitro experiments reveals that overexpression of miR-421 inhibited the TLR/MYD88 pathway. Besides, overexpressed miR-421 declined cell apoptosis but increased cell proliferation. It reveals that miR-421 targeting MYD88 could inhibit the apoptosis and autophagy of hippocampal neurons in epilepsy mice by down-regulating the TLR/MYD88 pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human MYD88
Informacje o cenach i dostępności nie są obecnie dostępne.