Merck
Pesticides in indoor and outdoor residential dust: a pilot study in a rural county of Taiwan.
Environmental science and pollution research international (2018-06-07)
Chien-Che Hung, Feng-Jung Huang, Ya-Qing Yang, Chia-Jung Hsieh, Chun-Chieh Tseng, Lih-Ming Yiin
ABSTRAKT
We conducted a pilot study to examine pesticides in dust of homes in a rural county of Taiwan. A total of 56 homes of pregnant women were included in the study. Indoor and outdoor dust was collected by a vacuum sampler and a dustpan/brush set, respectively. Nine pesticides were selected for analysis on gas chromatography-mass spectrometry with the detection limits being 0.088 ng/g or lower. The most detected pesticides were cypermethrin and chlorpyrifos, which appeared in 82.7 and 78.8% of indoor samples and 48.2 and 39.3% of outdoor samples, respectively. The detection of pesticides from indoor and outdoor dust, however, was not consistent, indicating different sources of pesticides. In addition to those two most detected, permethrin, prallethrin, and tetramethrin, which were common ingredients of insecticide products for indoor use, were also frequently found in indoor dust, suggesting that indoor use of such pesticide products may have been a major source. Fewer pesticides were found in outdoor dust, but the outdoor detection of chlorpyrifos was significantly associated with farms present inside the circles with radii of 50 and 100 m surrounding the homes (P = 0.021, 0.016). It is suggested that pesticide drift from agricultural areas to residential environments may have occurred. No seasonal effect on distribution of pesticides in dust was found, indicating that pesticides could be routinely used in Taiwan regardless of season. Compared with other international studies, this study shows relatively high levels of chlorpyrifos but low levels of pyrethroids (i.e., cypermethrin), reflecting a different pattern of pesticide use in Taiwan. Further studies need to be warranted for a better understanding of exposure to pesticides and the associated health effects.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Cyphenothrin, PESTANAL®, analytical standard, sum of isomers
Informacje o cenach i dostępności nie są obecnie dostępne.