Merck
TTK/hMps1 mediates the p53-dependent postmitotic checkpoint by phosphorylating p53 at Thr18.
Molecular and cellular biology (2009-04-01)
Yi-Fu Huang, Margaret Dah-Tsyr Chang, Sheau-Yann Shieh
ABSTRAKT
Upon prolonged arrest in mitosis, cells undergo adaptation and exit mitosis without cell division. These tetraploid cells are either eliminated by apoptosis or arrested in the subsequent G(1) phase in a spindle checkpoint- and p53-dependent manner. p53 has long been known to be activated by spindle poisons, such as nocodazole and Taxol, although the underlying mechanism remains elusive. Here we present evidence that stabilization and activation of p53 by spindle disruption requires the spindle checkpoint kinase TTK/hMps1. TTK/hMps1 phoshorylates the N-terminal domain of p53 at Thr18, and this phosphorylation disrupts the interaction with MDM2 and abrogates MDM2-mediated p53 ubiquitination. Phosphorylation at Thr18 enhances p53-dependent activation of not only p21 but also Lats2, two mediators of the postmitotic checkpoint. Furthermore, a phospho-mimicking substitution at Thr18 (T18D) is more competent than the phospho-deficient mutant (T18A) in rescuing the tetraploid checkpoint defect of p53-depleted cells. Our findings therefore provide a mechanism connecting the spindle checkpoint with p53 in the maintenance of genome stability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Informacje o cenach i dostępności nie są obecnie dostępne.