Merck
Cub domain-containing protein 1 negatively regulates TGF-β signaling and myofibroblast differentiation.
American journal of physiology. Lung cellular and molecular physiology (2018-01-20)
Nina Noskovičová, Katharina Heinzelmann, Gerald Burgstaller, Jürgen Behr, Oliver Eickelberg
ABSTRAKT
Fibroblasts are thought to be the prime cell type for producing and secreting extracellular matrix (ECM) proteins in the connective tissue. The profibrotic cytokine transforming growth factor-β1 (TGF-β1) activates and transdifferentiates fibroblasts into α-smooth muscle actin (α-SMA)-expressing myofibroblasts, which exhibit increased ECM secretion, in particular collagens. Little information, however, exists about cell-surface molecules on fibroblasts that mediate this transdifferentiation process. We recently identified, using unbiased cell-surface proteome analysis, Cub domain-containing protein 1 (CDCP1) to be strongly downregulated by TGF-β1. CDCP1 is a transmembrane glycoprotein, the expression and role of which has not been investigated in lung fibroblasts to date. Here, we characterized, in detail, the effect of TGF-β1 on CDCP1 expression and function, using immunofluorescence, FACS, immunoblotting, and siRNA-mediated knockdown of CDCP1. CDCP1 is present on interstitial fibroblasts, but not myofibroblasts, in the normal and idiopathic pulmonary fibrosis lung. In vitro, TGF-β1 decreased CDCP1 expression in a time-dependent manner by impacting mRNA and protein levels. Knockdown of CDCP1 enhanced a TGF-β1-mediated cell adhesion of fibroblasts. Importantly, CDCP1-depleted cells displayed an enhanced expression of profibrotic markers, such as collagen V or α-SMA, which was found to be independent of TGF-β1. Our data show, for the very first time that loss of CDCP1 contributes to fibroblast to myofibroblast differentiation via a potential negative feedback loop between CDCP1 expression and TGF-β1 stimulation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
MISSION® esiRNA, targeting human CDCP1
Informacje o cenach i dostępności nie są obecnie dostępne.