Merck
Photo and Thermo Dual-Responsive Copolymer Surfaces for Efficient Cell Capture and Release.
Chemphyschem : a European journal of chemical physics and physical chemistry (2018-03-16)
Yuwei Hao, Hongliang Liu, Guannan Li, Haijun Cui, Lei Jiang, Shutao Wang
ABSTRAKT
To overcome the low efficiency of single-responsive smart surfaces, we have constructed a dual-responsive smart surface - poly(spiropyran-co-N-isopropylacrylamide) (poly(SP-co-NiPAAm))-grafted silicon nanowire arrays - by combining photo-responsive SP and thermo-responsive NiPAAm units for enhancing the efficiencies of cancer-cell capture and release. These enhanced efficiencies probably originate from the binary cooperative effect of two responsive building units: NiPAAm units can decrease the steric hindrance between SP units during the isomerization while SP units can facilitate phase transition of NiPAAm units. This study provides a new strategy for designing smart materials and surfaces with efficient responsiveness for biomedical applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
Sigma-Aldrich
α-Bromoisobutyryl bromide, 98%
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)