Merck
CHCHD10 is involved in the development of Parkinson's disease caused by CHCHD2 loss-of-function mutation p.T61I.
Neurobiology of aging (2018-12-12)
Chengyuan Mao, Herui Wang, Haiyang Luo, Shuyu Zhang, Huisha Xu, Shuo Zhang, Jared Rosenblum, Zhilei Wang, Qi Zhang, Mibo Tang, Matthew J Shepard, Xiang Wang, Yaohe Wang, Zhengping Zhuang, Changhe Shi, Yuming Xu
ABSTRAKT
Previously we identified the p.Thr61Ile mutation in coiled-coil-helix-coiled-coil-helix domain containing 2 (CHCHD2) in a Chinese family with autosomal dominant Parkinson's disease. But the mechanism is still unclear. In this study, we explored the effects of CHCHD2 p.Thr61Ile mutation in cells and its association with coiled-coil-helix-coiled-coil-helix domain containing 10 (CHCHD10). We found that overexpression of Parkinson's disease-associated T61I mutant CHCHD2 did not produce mitochondrial dysfunction. Rather, its protective effect from stress was abrogated. And, the level of the CHCHD2 protein and mRNA in patient fibroblasts was not significantly different from control. In addition, CHCHD2 T61I mutation caused increased interaction with CHCHD10 and reduced CHCHD10 level. The mitochondrial ultrastructural alterations in CHCHD2 T61I mutant patient fibroblasts are similar to that of CHCHD10 mutations. We therefore propose that CHCHD10 is involved in the development of Parkinson's disease caused by CHCHD2 loss-of-function mutation p.T61I.