Merck
Influence of polar co-solutes and salt on the hydration of lipid membranes.
Physical chemistry chemical physics : PCCP (2019-07-26)
Amanuel Wolde-Kidan, Quoc Dat Pham, Alexander Schlaich, Philip Loche, Emma Sparr, Roland R Netz, Emanuel Schneck
ABSTRAKT
The influence of the co-solutes TMAO, urea, and NaCl on the hydration repulsion between lipid membranes is investigated in a combined experimental/simulation approach. Pressure-hydration curves obtained via sorption experiments reveal that the repulsion significantly increases when the membranes are loaded with co-solutes, most strongly for TMAO. As a result, the co-solutes retain additional water molecules and therefore provide membranes with a fluid and more physiological environment. The experimental data are quantitatively reproduced in complementary solvent-explicit atomistic molecular dynamics simulations, which yield the chemical potential of water. Simulation analysis reveals that the additional repulsion arises from the osmotic pressure generated by the co-solutes, an effect which is maximal for TMAO, due to its unfavorable interactions with the lipid headgroup layer and its extraordinarily high osmotic coefficient.