Merck
  • Whole-animal accumulation, oxidative stress, transcriptomic and metabolomic responses in the pink shrimp (Pandalus montagui) exposed to teflubenzuron.
Whole-animal accumulation, oxidative stress, transcriptomic and metabolomic responses in the pink shrimp (Pandalus montagui) exposed to teflubenzuron.
Journal of applied toxicology : JAT (2018-10-23)
Pål A Olsvik, Marte Aulin, Ole B Samuelsen, Rita Hannisdal, Ann-Lisbeth Agnalt, Bjørn T Lunestad
ABSTRAKT
The benzoylurea chitin synthesis inhibitor teflubenzuron, widely used against sea lice in North Atlantic aquaculture, may pose an environmental threat to non-targeted crustaceans. In this experiment, laboratory acclimated pink shrimp (Pandalus montagui), a species found in fjords with Atlantic salmon farming, were exposed to dietary teflubenzuron for 46 days (control; low dose: 0.01 μg/g; high dose: 0.1 μg/g). The exposure doses represent 0.1% and 1% of a standard treatment dose for Atlantic salmon. Mortality and prevalence of deformities, pharmacokinetics, oxidative stress and transcriptomic and metabolomic profiling were used to assess the response to teflubenzuron exposure. Mortality in the high-dose group was 25% (five of 20 individuals). No control or low-dose group shrimps died. Phenotypic responses,i.e., leg deformities (0 control, 6 low, 8 high) and cloudy eyes (0 control, 3 low, 7 high), were observed in some surviving shrimps (control n = 15, low n = 17, high n = 15). Accumulated levels of teflubenzuron in shrimps from the high-dose group ranged from 4.7 to 369 ng/g wet weight. Transcriptomic profiling showed very few significantly altered genes in the exposed shrimps. Teflubenzuron-induced changes to the metabolome pointed to well-known effects of benzoylurea agents, with reduced levels of N-acetylglucosamine indicating an effect on chitin synthesis. The metabolomic profiling showed that teflubenzuron exposure was associated with reduced energy metabolism. Some metabolites pointed to increased necrosis and/or bacterial overgrowth in the teflubenzuron-exposed shrimps. In conclusion, this study shows that teflubenzuron causes phenotypic effects in P. montagui exposed to 0.1% of the treatment dose given to Atlantic salmon.