Merck
Dual inhibition of Fes and Flt3 tyrosine kinases potently inhibits Flt3-ITD+ AML cell growth.
PloS one (2017-07-21)
Mark C Weir, Sabine Hellwig, Li Tan, Yao Liu, Nathanael S Gray, Thomas E Smithgall
ABSTRAKT
Acute myelogenous leukemia (AML) is often associated with activating mutations in the receptor tyrosine kinase, Flt3, including internal tandem duplications (ITDs) within the regulatory juxtamembrane region. Previous studies have linked Flt3-ITD to the activation of the Fes protein tyrosine kinase in AML, and RNAi-knockdown studies suggest that Fes may be required for Flt3 function. In this study, we tested Fes inhibitors from three different chemical classes for their growth-suppressive activity against Flt3-ITD+ myeloid leukemia cell lines (MV4-11, MOLM-13 and MOLM-14) vs. myeloid cells with wild-type Flt3 (THP-1). All Fes inhibitors selectively inhibited the growth of Flt3-ITD+ AML cells, with IC50 values for diaminopyrimidine and pyrrolopyridine inhibitors ranging from 19 to 166 nM. In contrast, a pyrazolopyrimidine inhibitor was less potent in Flt3-ITD+ AML cells, with IC50 values in the 1.0 μM range. In vitro kinase assays showed that the most potent inhibitors of Flt3-ITD+ AML cell proliferation blocked both Fes and Flt3-ITD kinase activity, while the pyrazolopyrimidine was more selective for Fes vs. Flt3-ITD. All three inhibitors induced significant apoptosis in Flt3-ITD+ AML cells, with potency equivalent to or greater than the established Flt3-ITD inhibitor, tandutinib. Transformation of TF-1 cells with Flt3-ITD resulted in constitutive activation of endogenous Fes, and rendered the cells highly sensitive to all three Fes inhibitors with IC50 values in the 30-500 nM range. The pyrrolopyridine compound also induced apoptotic responses in patient-derived Flt3-ITD+ AML bone marrow cells but not in normal bone marrow mononuclear cells. These results demonstrate that Fes kinase activity contributes to Flt3-ITD signaling in AML, and suggests that dual inhibition of both Flt3 and Fes may provide a therapeutic advantage for the treatment of Flt3-ITD+ AML.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tandutinib hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Przeciwciało przeciw fosfatydyloserynie, klon 1H6, Alexa Fluor 488, clone 1H6, Upstate®, from mouse