Merck
Measurement of autophagy in cells and tissues.
Methods in molecular biology (Clifton, N.J.) (2010-08-12)
Isei Tanida, Satoshi Waguri
ABSTRAKT
Two major proteolysis systems, the ubiquitin-proteasome system, and the autophagy-lysosome system, contribute to degradation of various types of protein and/or protein aggregates. In general, the autophagy-lysosome system is involved in bulk intracellular degradation of proteins and organelles, while the ubiquitin-proteasome system is selective. During autophagy, a cytosolic form of LC3 (LC3-I) is conjugated to phosphatidylethanolamine to form LC3-phosphatidylethanolamine conjugate (LC3-II), which is recruited to autophagosomal membranes, and LC3-II is degraded by lysosomal hydrolases after the fusion of autophagosomes with lysosomes. Therefore, lysosomal turnover of LC3-II reflects starvation-induced autophagic activity, and detection of LC3 by immunoblotting or immunofluorescence has become a reliable method for monitoring autophagy. When autophagy is impaired, the level of p62/SQSTM1, a ubiquitin- and LC3-binding protein, is increased in addition to the accumulation of ubiquitinated proteins. Here, we describe basic protocols to analyze endogenous LC3-II, p62, and autophagy-related proteins by immunoblotting, immunofluorescence, and electron microscopy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Informacje o cenach i dostępności nie są obecnie dostępne.