Merck
Fibulin-1c regulates transforming growth factor-β activation in pulmonary tissue fibrosis.
JCI insight (2019-07-26)
Gang Liu, Marion A Cooley, Andrew G Jarnicki, Theo Borghuis, Prema M Nair, Gavin Tjin, Alan C Hsu, Tatt Jhong Haw, Michael Fricker, Celeste L Harrison, Bernadette Jones, Nicole G Hansbro, Peter A Wark, Jay C Horvat, W Scott Argraves, Brian G Oliver, Darryl A Knight, Janette K Burgess, Philip M Hansbro
ABSTRAKT
Tissue remodeling/fibrosis is a major feature of all fibrotic diseases, including idiopathic pulmonary fibrosis (IPF). It is underpinned by accumulating extracellular matrix (ECM) proteins. Fibulin-1c (Fbln1c) is a matricellular ECM protein associated with lung fibrosis in both humans and mice, and stabilizes collagen formation. Here we discovered that Fbln1c was increased in the lung tissues of IPF patients and experimental bleomycin-induced pulmonary fibrosis. Fbln1c-deficient (-/-) mice had reduced pulmonary remodeling/fibrosis and improved lung function after bleomycin challenge. Fbln1c interacted with fibronectin, periostin and tenascin-c in collagen deposits following bleomycin challenge. In a novel mechanism of fibrosis Fbln1c bound to latent transforming growth factor (TGF)-β binding protein-1 (LTBP1) to induce TGF-β activation, and mediated downstream Smad3 phosphorylation/signaling. This process increased myofibroblast numbers and collagen deposition. Fbln1 and LTBP1 co-localized in lung tissues from IPF patients. Thus, Fbln1c may be a novel driver of TGF-β-induced fibrosis involving LTBP1 and may be an upstream therapeutic target.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-LTBP1 Antibody hinge region, from rabbit, purified by affinity chromatography
Informacje o cenach i dostępności nie są obecnie dostępne.