Merck
Insulin Signaling Regulates the FoxM1/PLK1/CENP-A Pathway to Promote Adaptive Pancreatic β Cell Proliferation.
Cell metabolism (2017-03-14)
Jun Shirakawa, Megan Fernandez, Tomozumi Takatani, Abdelfattah El Ouaamari, Prapaporn Jungtrakoon, Erin R Okawa, Wei Zhang, Peng Yi, Alessandro Doria, Rohit N Kulkarni
ABSTRAKT
Investigation of cell-cycle kinetics in mammalian pancreatic β cells has mostly focused on transition from the quiescent (G0) to G1 phase. Here, we report that centromere protein A (CENP-A), which is required for chromosome segregation during the M-phase, is necessary for adaptive β cell proliferation. Receptor-mediated insulin signaling promotes DNA-binding activity of FoxM1 to regulate expression of CENP-A and polo-like kinase-1 (PLK1) by modulating cyclin-dependent kinase-1/2. CENP-A deposition at the centromere is augmented by PLK1 to promote mitosis, while knocking down CENP-A limits β cell proliferation and survival. CENP-A deficiency in β cells leads to impaired adaptive proliferation in response to pregnancy, acute and chronic insulin resistance, and aging in mice. Insulin-stimulated CENP-A/PLK1 protein expression is blunted in islets from patients with type 2 diabetes. These data implicate the insulin-FoxM1/PLK1/CENP-A pathway-regulated mitotic cell-cycle progression as an essential component in the β cell adaptation to delay and/or prevent progression to diabetes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H3 (Ser10), marker mitozy, Upstate®, from rabbit
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)