Merck
Myxoid liposarcoma FUS-DDIT3 fusion oncogene induces C/EBP beta-mediated interleukin 6 expression.
International journal of cancer (2005-02-03)
Melker Göransson, Erik Elias, Anders Ståhlberg, Anita Olofsson, Carola Andersson, Pierre Aman
ABSTRAKT
The myxoid/round cell liposarcoma oncogene FUS-DDIT3 is the result of a translocation derived gene fusion between the splicing factor FUS and DDIT3. In order to investigate the downstream targets of DDIT3, and the transforming effects of the FUS-DDIT3 fusion protein, we have introduced DDIT3-GFP and FUS-DDIT3-GFP constructs into a human fibrosarcoma cell line. The gene expression profiles of stable transfectants were compared to the original fibrosarcoma cell line by microarray analysis. We here report that the NFkappaB and C/EBP beta controlled gene IL6 is upregulated in DDIT3- and FUS-DDIT3-expressing fibrosarcoma cell lines and in myxoid liposarcoma cell lines. Strong expression of the tumor associated multifunctional cytokine interleukin 6 was confirmed both at mRNA and protein level. Knockdown experiments using siRNA against CEBPB transcripts showed that the effect of FUS-DDIT3 on IL6 expression is C/EBP beta dependent. Chromatin immunoprecipitation revealed direct interaction between the IL6 promoter and the C/EBP beta protein. In addition, the effect of DDIT3 and FUS-DDIT3 on the expression of other acute phase genes was examined using real-time PCR. We demonstrate for the first time that DDIT3 and FUS-DDIT3 show opposite transcriptional regulation of IL8 and suggest that FUS-DDIT3 may affect the synergistic activation of promoters regulated by C/EBP beta and NFkappaB.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-acetylo-histonowe H4, serum, Upstate®
Sigma-Aldrich
Zestaw do immunoprecypitacji acetylo-histonu H4 (ChIP), Acetyl-Histone H4 Immunoprecipitation (ChIP) Assay Kit used to immunoprecipitate transcriptionally active chromatin from mammalian cells using anti-Acetyl-Histone H4, ChIP grade rabbit antiserum.