Merck
Cell-autonomous and non-cell autonomous effects of neuronal BIN1 loss in vivo.
PloS one (2019-08-14)
Kathleen M McAvoy, Hameetha Rajamohamed Sait, Galina Marsh, Michael Peterson, Taylor L Reynolds, Jake Gagnon, Sarah Geisler, Prescott Leach, Chris Roberts, Ellen Cahir-McFarland, Richard M Ransohoff, Andrea Crotti
ABSTRAKT
BIN1 is the most important risk locus for Late Onset Alzheimer's Disease (LOAD), after ApoE. BIN1 AD-associated SNPs correlate with Tau deposition as well as with brain atrophy. Furthermore, the level of neuronal-specific BIN1 isoform 1 protein is decreased in sporadic AD cases in parallel with neuronal loss, despite an overall increase in BIN1 total mRNA. To address the relationship between reduction of BIN1 and neuronal cell loss in the context of Tau pathology, we knocked-down endogenous murine Bin1 via stereotaxic injection of AAV-Bin1 shRNA in the hippocampus of mice expressing Tau P301S (PS19). We observed a statistically significant reduction in the number of neurons in the hippocampus of mice injected with AAV-Bin1 shRNA in comparison with mice injected with AAV control. To investigate whether neuronal loss is due to deletion of Bin1 selectively in neurons in presence Tau P301S, we bred Bin1flox/flox with Thy1-Cre and subsequently with PS19 mice. Mice lacking neuronal Bin1 and expressing Tau P301S showed increased mortality, without increased neuropathology, when compared to neuronal Bin1 and Tau P301S-expressing mice. The loss of Bin1 isoform 1 resulted in reduced excitability in primary neurons in vitro, reduced neuronal c-fos expression as well as in altered microglia transcriptome in vivo. Taken together, our data suggest that the contribution of genetic variation in BIN1 locus to AD risk could result from a cell-autonomous reduction of neuronal excitability due to Bin1 decrease, exacerbated by the presence of aggregated Tau, coupled with a non-cell autonomous microglia activation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-NeuN Antibody, serum, from guinea pig
Informacje o cenach i dostępności nie są obecnie dostępne.
Millipore
Accutase cell detachment solution, A cell detachment solution of proteolytic & collagenolytic enzymes. The reagent is useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Informacje o cenach i dostępności nie są obecnie dostępne.
Roche
cOmplete, Mini, Koktajl inhibitorów proteaz bez EDTA, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, ≥97.0% (AT)
Informacje o cenach i dostępności nie są obecnie dostępne.