Merck
Expression of lipid metabolism genes provides new insights into intramuscular fat deposition in Laiwu pigs.
Asian-Australasian journal of animal sciences (2019-09-05)
Hui Wang, Jin Wang, Dan-Dan Yang, Zong-Li Liu, Yong-Qing Zeng, Wei Chen
ABSTRAKT
The objective of this study was to measure the special expression pattern of lipid metabolism genes and investigate the molecular mechanisms underlying intramuscular fat (IMF) deposition in Longissimus dorsi muscle of Laiwu pigs. Thirty-six pigs (Laiwu n = 18; Duroc×Landrace×Yorkshire n = 18) were used for the measurement of the backfat thickness, marbling score, IMF content, and expression of lipid metabolism genes. Significant correlations were found between IMF content and the mRNA expression of lipid metabolism genes. Of the 14 fat deposition genes measured, fatty acid synthase (FASN) showed the strongest correlation (r = 0.75, p = 0.001) with IMF content, and of the 6 fat removal genes, carnitine palmitoyl transferase 1B (CPT1B) exhibited the greatest negative correlation (r = -0.66, p = 0.003) with IMF content in Laiwu pig. Multiple regression analysis showed that CPT1B, FASN, solute carrier family 27 member 1 (SLC27A1), and fatty acid binding protein 3 (FABP3) contributed 38% of the prediction value for IMF content in Laiwu pigs. Of these four variables, CPT1B had the greatest contribution to IMF content (14%) followed by FASN (11%), SLC27A1 (9%), and FABP3 (4%). Our results indicate that the combined effects of an upregulation in fat deposition genes and downregulation in fat removal genes promotes IMF deposition in Laiwu pigs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.