Merck
  • Encapsulation of the black pepper (Piper nigrum L.) essential oil by lactoferrin-sodium alginate complex coacervates: Structural characterization and simulated gastrointestinal conditions.
Encapsulation of the black pepper (Piper nigrum L.) essential oil by lactoferrin-sodium alginate complex coacervates: Structural characterization and simulated gastrointestinal conditions.
Food chemistry (2020-02-12)
Lívia Pinto Heckert Bastos, Carlos Henrique Corrêa Dos Santos, Mario Geraldo de Carvalho, Edwin Elard Garcia-Rojas
ABSTRAKT
Black pepper essential oil (EO) was encapsulated by complex coacervation with lactoferrin and sodium alginate using transglutaminase as a cross-linking agent. The encapsulation efficiency varied from 31.66 to 84.48%. Chemical and morphological characteristics suggest that the EO was encapsulated in a lactoferrin/sodium alginate shell. The chemical composition of the encapsulated EO was identified by gas chromatography (GC) and nuclear magnetic resonance (NMR). The GC and NMR analyses indicated good core protection with the materials used. The stability of the black pepper EO capsules under in vitro digestion was evaluated. Theses capsules demonstrated the low release of the EO during gastric digestion and higher release in intestinal digestion. These results suggest that these capsules can be used to transport active ingredients and that they are resistant to oral and gastric conditions that were tested in vitro.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications
Millipore
Bile bovine, dried, unfractionated
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein