Merck
  • Vasodilation of rat skeletal muscle arteries by the novel BK channel opener GoSlo is mediated by the simultaneous activation of BK and Kv 7 channels.
Vasodilation of rat skeletal muscle arteries by the novel BK channel opener GoSlo is mediated by the simultaneous activation of BK and Kv 7 channels.
British journal of pharmacology (2019-10-29)
Olga Zavaritskaya, Srikanth Dudem, Dongyu Ma, Kaneez E Rabab, Sarah Albrecht, Dmitry Tsvetkov, Mario Kassmann, Keith Thornbury, Mitko Mladenov, Claire Kammermeier, Gerard Sergeant, Nicholas Mullins, Ornella Wouappi, Hannah Wurm, Aimo Kannt, Maik Gollasch, Mark A Hollywood, Rudolf Schubert
ABSTRAKT
BK channels play important roles in various physiological and pathophysiological processes and thus have been the target of several drug development programmes focused on creating new efficacious BK channel openers, such as the GoSlo-SR compounds. However, the effect of GoSlo-SR compounds on vascular smooth muscle has not been studied. Therefore, we tested the hypothesis that GoSlo-SR compounds dilate arteries exclusively by activating BK channels. Experiments were performed on rat Gracilis muscle, saphenous, mesenteric and tail arteries using isobaric and isometric myography, sharp microelectrodes, digital droplet PCR and the patch-clamp technique. GoSlo-SR compounds dilated isobaric and relaxed and hyperpolarised isometric vessel preparations and their effects were abolished after (a) functionally eliminating K+ channels by pre-constriction with 50 mM KCl or (b) blocking all K+ channels known to be expressed in vascular smooth muscle. However, these effects were not blocked when BK channels were inhibited. Surprisingly, the Kv 7 channel inhibitor XE991 reduced their effects considerably, but neither Kv 1 nor Kv 2 channel blockers altered the inhibitory effects of GoSlo-SR. However, the combined blockade of BK and Kv 7 channels abolished the GoSlo-SR-induced relaxation. GoSlo-SR compounds also activated Kv 7.4 and Kv 7.5 channels expressed in HEK 293 cells. This study shows that GoSlo-SR compounds are effective relaxants in vascular smooth muscle and mediate their effects by a combined activation of BK and Kv 7.4/Kv 7.5 channels. Activation of Kv 1, Kv 2 or Kv 7.1 channels or other vasodilator pathways seems not to be involved.