Merck
The expression, purification, and substrate analysis of matrix metalloproteinases in Drosophila melanogaster.
Protein expression and purification (2020-03-24)
Di Wen, Zhi Chen, Zeyan Zhang, Qiangqiang Jia
ABSTRAKT
Matrix metalloproteinases (MMPs) are evolutionarily conserved extracellular matrix proteinases. Genetic analysis of the Drosophila MMPs, Mmp1 and Mmp2, in vivo reveal that they play vital roles in tissue remodeling. Although the catalytic domain (CD) undertakes most MMP functions, few studies have sought to demonstrate the biochemical properties of the CDs of fly MMPs. Here, we identified the overexpression, purification, and refolding of the CDs of Drosophila Mmp1 and Mmp2 for biochemical studies. Zymography assays and substrate degradation analysis showed that both Mmp1-CD and Mmp2-CD were able to digest casein, gelatin, fibronectin, collagen (types I, IV, and V), while Mmp2-CD showed much higher degradation activity compared with Mmp1-CD. Moreover, human collagen III could be degraded by Mmp1-CD but not Mmp2-CD, and rat collagen I and laminin could be degraded by Mmp2-CD but not Mmp1-CD, suggesting that Drosophila Mmp1 and Mmp2 might have overlapping yet distinct substrate specificity. Using synthetic fluorescent substrates, we further demonstrated that the enzymatic activity of Mmp1-CD and Mmp2-CD could be inhibited by human tissue inhibitors of metalloproteinases (TIMPs). These results reveal the context of the cooperative yet distinct roles of Mmp1 and Mmp2 in tissue remodeling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
TIMP-1 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
TIMP-2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Casein solution from bovine milk, 5% in water
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder