Merck
Particle Monolayer-Stabilized Light-Sensitive Liquid Marbles from Polypyrrole-Coated Microparticles.
Langmuir : the ACS journal of surfaces and colloids (2020-02-23)
Yuta Asaumi, Marcel Rey, Nicolas Vogel, Yoshinobu Nakamura, Syuji Fujii
ABSTRAKT
Liquid marbles are water droplets coated with solid particles that prevent coalescence and allow storage, transport, and handling of liquids in the form of a powder. Here, we report on the formation of liquid marbles that are stabilized entirely by a single monolayer of solid particles and thus minimize the amount of required solid material. As stabilizing particles, we synthesize relatively monodisperse, 80 μm-sized polystyrene (PS) particles coated with heptadecafluorooctanesulfonic acid-doped polypyrrole (PPy-C8F) shell (PS/PPy-C8F particles) by aqueous chemical oxidative seeded polymerization of pyrrole using FeCl3 as an oxidant and heptadecafluorooctanesulfonic acid as a hydrophobic dopant. We characterize the physicochemical properties of the particles as a function of the PPy-C8F loading. Laser diffraction particle size analyses of dilute aqueous suspensions indicate that the polymer particles disperse stably in water medium before and after coating with the PPy-C8F shell. X-ray photoelectron spectroscopy studies indicate the formation of a PPy-C8F shell around the PS seed particles, which was further supported by deflated morphologies observed by scanning electron microscopy after extraction of the PS component from the PS/PPy-C8F particles. We investigate the performance of the dried PS/PPy-C8F particles to stabilize liquid marbles. Stereo- and laser microscope observations, as well as gravimetric studies, confirm that the PS/PPy-C8F particles adsorb to the water droplet surface in the form of a particle monolayer with the characteristic hexagonal close-packed structure expected for spherical (colloidal) particles. Mechanical integrity of the liquid marble increases with an increase of PPy-C8F loading of the PS/PPy-C8F particles. The water contact angle of the PS/PPy-C8F particles at air-water interface increases from 82 ± 12° to 102 ± 17° with an increase of PPy-C8F loading. This increase in water contact angle directly correlates with the shape of the LM, with higher contact angles giving more spherical LMs. Furthermore, the broadband light absorption properties of the PPy coating was used to control evaporation rate of the enclosed water phase on demand by irradiation with a near-infrared laser. The evaporation rate could be finely controlled by the thickness of the PPy-C8F shell of the particles stabilizing the liquid marbles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl 2-cyanoacrylate, liquid
Informacje o cenach i dostępności nie są obecnie dostępne.