Merck
Intermediate progenitors support migration of neural stem cells into dentate gyrus outer neurogenic niches.
eLife (2020-04-03)
Branden R Nelson, Rebecca D Hodge, Ray Am Daza, Prem Prakash Tripathi, Sebastian J Arnold, Kathleen J Millen, Robert F Hevner
ABSTRAKT
The hippocampal dentate gyrus (DG) is a unique brain region maintaining neural stem cells (NCSs) and neurogenesis into adulthood. We used multiphoton imaging to visualize genetically defined progenitor subpopulations in live slices across key stages of mouse DG development, testing decades old static models of DG formation with molecular identification, genetic-lineage tracing, and mutant analyses. We found novel progenitor migrations, timings, dynamic cell-cell interactions, signaling activities, and routes underlie mosaic DG formation. Intermediate progenitors (IPs, Tbr2+) pioneered migrations, supporting and guiding later emigrating NSCs (Sox9+) through multiple transient zones prior to converging at the nascent outer adult niche in a dynamic settling process, generating all prenatal and postnatal granule neurons in defined spatiotemporal order. IPs (Dll1+) extensively targeted contacts to mitotic NSCs (Notch active), revealing a substrate for cell-cell contact support during migrations, a developmental feature maintained in adults. Mouse DG formation shares conserved features of human neocortical expansion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Prospero homeobox protein 1/PROX1 Antibody, from rabbit, purified by affinity chromatography
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Tamoxifen, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.