Merck
Tilt-table Echocardiography Unmasks Early Diastolic Dysfunction in Patients With Hemoglobinopathies.
Journal of pediatric hematology/oncology (2020-04-15)
Kandice Mah, Aisha Bruce, Norazah Zahari, Mary Anne Venner, Kelvin Chow, Richard B Thompson, Nee S Khoo, Edythe B Tham
ABSTRAKT
Individuals with hemoglobinopathy (sickle cell anemia and thalassemia major) are at risk for cardiac complications such as heart failure and cardiomyopathy. Diastolic dysfunction is known to precede systolic dysfunction in many cardiac diseases. This study sought to determine whether changes in left atrial (LA) function during manipulation of cardiac preload by tilt-table echocardiography can unmask subclinical diastolic dysfunction in pediatric patients with hemoglobinopathies. Eleven sickle cell anemia, 9 transfusion-dependent thalassemia major, and 10 control subjects underwent tilt-table echocardiogram in the supine (loading) and 30-degree upright (unloading) positions and cardiac magnetic resonance imaging (MRI). Echocardiography assessed LA and left ventricular (LV) strain, strain rate, mitral inflow, and annular velocities. MRI assessed LV function, myocardial T1 and T2* for iron deposition. Both thalassemia major and sickle cell anemia patients had normal LV function and no evidence of cardiac iron deposition on MRI T2* measurements. During cardiac loading, controls appropriately increased LA conduit (P=0.002) and reservoir strain (P=0.002), mitral e' velocity (P<0.0001) and medial e' velocity (P=0.002), while the hemoglobinopathy patients showed no change in these parameters. In pediatric sickle cell anemia and thalassemia, tilt-table echocardiography unmasked a failure to augment LA function in response to loading, suggesting altered myocardial relaxation is present, before evidence of iron overload or systolic dysfunction.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Deferasirox, ≥98% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.