Merck
FBXO11/PRMT9, a new protein arginine methyltransferase, symmetrically dimethylates arginine residues.
Biochemical and biophysical research communications (2006-02-21)
Jeffry R Cook, Jin-Hyung Lee, Zhi-Hong Yang, Christopher D Krause, Nicole Herth, Ralf Hoffmann, Sidney Pestka
ABSTRAKT
We have identified a protein, FLJ12673 or FBXO11, that contains domains characteristically present in protein arginine methyltransferases (PRMTs). Immuno-purified protein expressed from one of the four splice variants in HeLa cells and in Escherichia coli exhibited methyltransferase activity. Monomethylarginine, symmetric, and asymmetric dimethylarginine (SDMA, ADMA) were formed on arginine residues. Accordingly, we have designated the protein PRMT9. PRMT9 is the third member of the PRMT family that forms SDMA modifications in proteins. Structurally, this protein is distinct from all other known PRMTs implying that convergent evolution allowed this protein to develop the ability to methylate arginine residues and evolved elements conserved in PRMTs to accomplish this.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PRMT9 human, recombinant, expressed in baculovirus infected Sf9 cells, ≥30% (SDS-PAGE)
Informacje o cenach i dostępności nie są obecnie dostępne.