Merck
Postnatal, ontogenic liver growth accomplished by biliary/oval cell proliferation and differentiation.
PloS one (2020-05-30)
Armanda Szücs, Sándor Paku, Endre Sebestyén, Péter Nagy, Katalin Dezső
ABSTRAKT
The liver is well known for its enormous regenerative capacity. If the hepatocytes are compromised the reserve stem cells can regrow the lost tissue by means of oval cells differentiating into hepatocytes. We were curious whether this standby system was able to compensate for ontogenic liver growth arrested by 2-acetylaminofluorene (AAF) treatment or if it can be influenced by cholic acid, known to promote liver growth in several reactions. (i) Four weeks-old (60-70g) male F344 rats were kept on standard chow and treated with solvent only, (ii) others were kept on 0,2% cholic acid (CA) enriched diet, (iii) treated with AAF, or (iiii) given a combination of CA diet and AAF treatment (AAF/CA). The proliferative response of epithelial cells was characterized by pulse bromodeoxyuridine labelling. The relative gene expression levels of senescence-related factors and bile acid receptors were determined by quantitative real-time polymerase chain reaction analysis. AAF administration efficiently inhibited the physiological proliferation of hepatocytes in young, male F344 rats after weaning. The activation of stem cells was indicated by the increased proliferation of periportal biliary/oval cells (B/OC). If the rats were fed additionally by cholic acid enriched diet, typical oval cell reaction emerged, subsequently the oval cells differentiated into hepatocytes restituting liver growth. This reaction was mediated by increased production of HGF, IL-6 and SCF by the damaged liver. Moreover, upregulation of FXR expression on B/OC made them competent for bile acids. Our results indicate that endogenous, autocrine mechanisms involved in liver ontogeny are also able to activate the backup regenerative machinery of stem cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining