Merck
VPS72/YL1-Mediated H2A.Z Deposition Is Required for Nuclear Reassembly after Mitosis.
Cells (2020-07-28)
Daniel Moreno-Andrés, Hideki Yokoyama, Anja Scheufen, Guillaume Holzer, Hongqi Lue, Anna Katharina Schellhaus, Marion Weberruss, Masatoshi Takagi, Wolfram Antonin
ABSTRAKT
The eukaryotic nucleus remodels extensively during mitosis. Upon mitotic entry, the nuclear envelope breaks down and chromosomes condense into rod-shaped bodies, which are captured by the spindle apparatus and segregated during anaphase. Through telophase, chromosomes decondense and the nuclear envelope reassembles, leading to a functional interphase nucleus. While the molecular processes occurring in early mitosis are intensively investigated, our knowledge about molecular mechanisms of nuclear reassembly is rather limited. Using cell free and cellular assays, we identify the histone variant H2A.Z and its chaperone VPS72/YL1 as important factors for reassembly of a functional nucleus after mitosis. Live-cell imaging shows that siRNA-mediated downregulation of VPS72 extends the telophase in HeLa cells. In vitro, depletion of VPS72 or H2A.Z results in malformed and nonfunctional nuclei. VPS72 is part of two chromatin-remodeling complexes, SRCAP and EP400. Dissecting the mechanism of nuclear reformation using cell-free assays, we, however, show that VPS72 functions outside of the SRCAP and EP400 remodeling complexes to deposit H2A.Z, which in turn is crucial for formation of a functional nucleus.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human VPS72
Sigma-Aldrich
Anti-Pontin antibody, Mouse monoclonal, clone 5G3-11, purified from hybridoma cell culture
Sigma-Aldrich
Przeciwciało anty-histonowe H2A.Z, serum, Upstate®
Sigma-Aldrich
Przeciwciało przeciw trimetylo-histonowi H3 (Lys9), serum, Upstate®
Sigma-Aldrich
5-Ethynyl uridine, ≥95%
Sigma-Aldrich
Anti-Ki-67 Antibody, clone Ki-S5, clone Ki-S5, Chemicon®, from mouse