Merck
Passive diffusion through nuclear pore complexes regulates levels of the yeast SAGA and SLIK coactivator complexes.
Journal of cell science (2020-02-14)
Tadashi Makio, Richard W Wozniak
ABSTRAKT
Nuclear pore complexes (NPCs) control gene expression by regulating the bi-directional exchange of proteins and RNAs between nuclear and cytoplasmic compartments, including access of transcriptional regulators to the nucleoplasm. Here, we show that the yeast (Saccharomyces cerevisiae) nucleoporin Nup170, in addition to binding and silencing subtelomeric genes, supports transcription of genes regulated by the SAGA transcriptional activator complex. Specifically, we show that a lower amount of SAGA complex is bound to target genes in the absence of Nup170. Consistent with this observation, levels of the SAGA complex are decreased in cells lacking Nup170, while those of the SAGA-related SLIK complexes are increased. This change in the ratio of SAGA to SLIK complexes is due to increased nuclear activity of Pep4, a protease responsible for production of the SLIK complex. Further analyses of various nucleoporin mutants revealed that the increased nuclear entry of Pep4 observed in the nup170Δ mutant likely occurs as the consequence of an increase in the sieving limits of the NPC diffusion channel. On the basis of these results, we propose that changes in passive diffusion rates represent a mechanism for regulating SAGA- and SLIK complex-mediated transcriptional events.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Protein A antibody produced in rabbit, fractionated antiserum, lyophilized powder
Roche
Anty-GFP, from mouse IgG1κ (clones 7.1 and 13.1)