Merck
Bioassay guided purification of the antimicrobial fraction of a Brazilian propolis from Bahia state.
BMC complementary and alternative medicine (2009-08-01)
Myrella Lessio Castro, Walquíria Ribeiro Vilela, Rogéria Cristina Zauli, Masaharu Ikegaki, Vera Lúcia Garcia Rehder, Mary Ann Foglio, Severino Matias de Alencar, Pedro Luiz Rosalen
ABSTRAKT
Brazilian propolis type 6 (Atlantic forest, Bahia) is distinct from the other types of propolis especially due to absence of flavonoids and presence of other non-polar, long chain compounds, but presenting good in vitro and in vivo antimicrobial activity. Several authors have suggested that fatty acids found in this propolis might be responsible for its antimicrobial activity; however, so far no evidence concerning this finding has been reported in the literature. The goals of this study were to evaluate the antibacterial activity of the main pure fatty acids in the ethanolic extract and fractions and elucidate the chemical nature of the bioactive compounds isolated from Brazilian propolis type 6. Brazilian propolis type 6 ethanolic extract (EEP), hexane fraction (H-Fr), major fatty acids, and isolated sub-fractions were analyzed using high performance liquid chromatography (HPLC), high resolution gas chromatography with flame ionization detection (HRGC-FID), and gas chromatography-mass spectrometry (GC-MS). Three sub-fractions of H-Fr were obtained through preparative HPLC. Antimicrobial activity of EEP, H-Fr, sub-fractions, and fatty acids were tested against Staphyloccus aureus ATCC 25923 and Streptococcus mutans Ingbritt 1600 using minimum inhibitory concentration (MIC) and minimum bactericidal concentration (MBC). EEP and H-Fr inhibited the growth of the microorganisms tested; nevertheless, no antimicrobial activity was found for the major fatty acids. The three sub-fractions (1, 2, and 3) were isolated from H-Fr by preparative HPLC and only sub-fraction 1 showed antimicrobial activity. a) The major fatty acids tested were not responsible for the antimicrobial activity of propolis type 6; b) Sub-fraction 1, belonging to the benzophenone class, was responsible for the antimicrobial activity observed in the present study. The identification of the bioactive compound will improve the development of more efficient uses of this natural product.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Chlorhexidine digluconate solution, 20% in H2O
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture