Merck
LDHA-mediated ROS generation in chondrocytes is a potential therapeutic target for osteoarthritis.
Nature communications (2020-07-11)
Manoj Arra, Gaurav Swarnkar, Ke Ke, Jesse E Otero, Jun Ying, Xin Duan, Takashi Maruyama, Muhammad Farooq Rai, Regis J O'Keefe, Gabriel Mbalaviele, Jie Shen, Yousef Abu-Amer
ABSTRAKT
The contribution of inflammation to the chronic joint disease osteoarthritis (OA) is unclear, and this lack of clarity is detrimental to efforts to identify therapeutic targets. Here we show that chondrocytes under inflammatory conditions undergo a metabolic shift that is regulated by NF-κB activation, leading to reprogramming of cell metabolism towards glycolysis and lactate dehydrogenase A (LDHA). Inflammation and metabolism can reciprocally modulate each other to regulate cartilage degradation. LDHA binds to NADH and promotes reactive oxygen species (ROS) to induce catabolic changes through stabilization of IκB-ζ, a critical pro-inflammatory mediator in chondrocytes. IκB-ζ is regulated bi-modally at the stages of transcription and protein degradation. Overall, this work highlights the function of NF-κB activity in the OA joint as well as a ROS promoting function for LDHA and identifies LDHA as a potential therapeutic target for OA treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6-Aminonicotinamide, 99%
Sigma-Aldrich
Dioctan 2′,7′-dichlorodihydrofluoresceiny, ≥97%
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sigma-Aldrich
Dihydroethidium, ≥95%
Roche
Collagenase D, from Clostridium histolyticum
Sigma-Aldrich
SC-514, ≥97% (HPLC)
Sigma-Aldrich
VAS2870, ≥97% (HPLC)
Sigma-Aldrich
Polybrene Infection / Transfection Reagent, A highly efficient method of gene transfer into mammalian cells leveraging infection with retroviral vectors.
Sigma-Aldrich
Sodium oxamate, ≥98%