Merck
A Concise, Modular Antibody-Oligonucleotide Conjugation Strategy Based on Disuccinimidyl Ester Activation Chemistry.
Chembiochem : a European journal of chemical biology (2019-02-16)
Gang Li, Raymond E Moellering
ABSTRAKT
The synthesis of antibody-oligonucleotide conjugates has enabled the development of highly sensitive bioassays for specific epitopes in the laboratory and clinic. Most synthetic schemes to generate these hybrid molecules require expensive reagents, significant quantities of input antibody, and multistep purification routes; thus limiting widespread application. Herein a facile and robust conjugation strategy is reported that involves "plug-and-play" antibody conjugation with succinimidyl-functionalized oligonucleotides, which are high yielding and compatible for use directly after buffer exchange. The succinimidyl-linked oligonucleotides are synthesized with 5'-amine-modified oligonucleotides and disuccinimidyl suberate (DSS), both of which are inexpensive and commercially available. Direct incubation of the resulting stable succinimidyl- oligonucleotide conjugates with commercial antibodies yields conjugates ready for use after benchtop buffer exchange. It is demonstrated that the resulting oligonucleotide-antibody and oligonucleotide-streptavidin conjugates retain potent and specific binding in activity-dependent proximity ligation imaging, and proximity ligation-mediated qPCR detection of endogenous proteins in native cellular contexts down to picogram levels of whole proteome. This DSS conjugation strategy should be widely applicable in the synthesis of protein-oligonucleotide conjugates.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Przeciwciało anty-Myc Tag, klon 4A6, clone 4A6, Upstate®, from mouse
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Informacje o cenach i dostępności nie są obecnie dostępne.