Merck
Human triple cell co-culture for evaluation of bone implant materials.
Integrative biology : quantitative biosciences from nano to macro (2013-02-20)
Frederik Wein, Arie Bruinink
ABSTRAKT
Central to the formation of tissue at implant surfaces are the interactions between multiple cell types including fibroblasts, endothelial cells and, in the case of bone, cells of the osteoblastic lineage. To date the importance of population dynamics and interactions have been largely neglected in the in vitro evaluation of biomaterials. To fill this gap we have developed a co-culture system using 3 cell types, primary human bone marrow stromal cells (HBMC), microvascular endothelial cells (HMVEC) and abdominal dermal fibroblasts (HDF). Proliferation of each cell type separately and differentiation of HBMC were determined by flow cytometry analysis. The medium used promoted HBMC differentiation toward osteoblasts without affecting the state of differentiation of HDF and HMVEC. Furthermore, HBMC are strongly affected by HDF and HMVEC, and vice versa. When used on a titanium coated substrate the triple cell culture system identified preferential HBMC proliferation relative to HDF if HMVEC was present. This developed culture system represents a new, optimised and potentially predictive approach to evaluate biomaterial biocompatibility early in development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-CD31(PECAM-1) antibody , Mouse monoclonal, clone WM-59, purified from hybridoma cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse, clone KDR-1, ascites fluid
Informacje o cenach i dostępności nie są obecnie dostępne.