Merck
Study on synthesis and biological activity of a galactosylated piperazinyl porphyrin.
Bioorganic & medicinal chemistry letters (2006-09-26)
He-Ping Li
ABSTRAKT
In order to obtain an carcinoma-selective drug, the synthesis and characterization of 5,10,15,20-tetra[4-(4'-galactosylpiperazinyl)phenyl]porphyrin (TGPP) is reported. The biological activity on cancer cells and the pharmacokinetics are also reported as preliminary results showing a very high liver to skin ratio and short retention time in tissues, and thus promising activity in photodynamic therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder