Merck
  • Cytarabine nanotherapeutics with increased stability and enhanced lymphoma uptake for tailored highly effective therapy of mantle cell lymphoma.
Cytarabine nanotherapeutics with increased stability and enhanced lymphoma uptake for tailored highly effective therapy of mantle cell lymphoma.
Acta biomaterialia (2020-11-14)
Robert Pola, Eva Pokorná, Petra Vočková, Eliška Böhmová, Michal Pechar, Jana Karolová, Jan Pankrác, Luděk Šefc, Karel Helman, Marek Trněný, Tomáš Etrych, Pavel Klener
ABSTRAKT
Mantle cell lymphoma (MCL) is a rare subtype of B-cell non-Hodgkin lymphoma (B-NHL) with chronically relapsing clinical course. Implementation of cytarabine (araC) into induction and salvage regimen became standard of care for majority of MCL patients. In this study, tailored N-(2-hydroxypropyl)methacrylamide (HPMA)-based polymer nanotherapeutics containing covalently bound araC (araC co-polymers) were designed, synthesized and evaluated for their anti-lymphoma efficacy in vivo using a panel of six patient-derived lymphoma xenografts (PDX) derived from newly diagnosed and relapsed / refractory (R/R) MCL. While free araC led to temporary inhibition of growth of MCL tumors, araC co-polymers induced long-term disappearance of the engrafted lymphomas with no observed toxicity even in the case of PDX models derived from patients, who relapsed after high-dose araC-based treatments. The results provide sound preclinical rationale for the use of HPMA-based araC co-polymers in induction, salvage or palliative therapy of MCL patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
2-(Dimethylamino)pyridine, 97%
Informacje o cenach i dostępności nie są obecnie dostępne.